Anti-GOSR1 Mouse Polyclonal Antibody
Catalog # ABNOH00009527-B01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Golgi SNAP receptor complex member 1
- Antigen symbol:GOSR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9527
- Antigen synonyms:P28|GOLIM2|GOS28/P28|GOS-28|GOS28|GS28
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GOSR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human