Anti-SNAP29 Rabbit Polyclonal Antibody
Catalog # ABNOH00009342-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Synaptosomal-Associated Protein 29kDa
- Antigen symbol:SNAP29
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9342
- Antigen synonyms:SNAP-29|Gs32|Vesicle-membrane fusion protein|Golgi SNARE of 32 kDa|Soluble 29 kDa NSF attachment protein|SNAP29
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SNAP29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat