Anti-LATS1 Mouse Monoclonal Antibody [clone: 3A7]
Catalog # ABNOH00009113-M09
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Large tumor suppressor kinase 1
- Antigen symbol:LATS1
- Clonality:Monoclonal
- Clone:3A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9113
- Antigen synonyms:WARTS|Large tumor suppressor homolog 1|LATS1_HUMAN.|WARTS protein kinase|Serine threonine protein kinase LATS1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: LATS1
Clonality: Monoclonal
Clone: 3A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human