Anti-FGF18 Mouse Polyclonal Antibody
Catalog # ABNOH00008817-B01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast growth factor 18
- Antigen symbol:FGF18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8817
- Antigen synonyms:ZFGF5|FGF-18
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: FGF18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human