Anti-SNTA1 Mouse Polyclonal Antibody
Catalog # ABNOH00006640-B01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Syntrophin alpha 1
- Antigen symbol:SNTA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6640
- Antigen synonyms:TACIP1|SNT1|LQT12|dJ1187J4.5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SNTA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human