Anti-RPS4X Mouse Monoclonal Antibody [clone: 3E10]
Catalog # ABNOH00006191-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal Protein S4, X-linked
- Antigen symbol:RPS4X
- Clonality:Monoclonal
- Clone:3E10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6191
- Antigen synonyms:cell cycle gene 2|X isoform|40S ribosomal protein S4|ribosomal protein S4X isoform|SCAR|RPS4|CCG2|single-copy abundant mRNA|SCR10|single copy abundant mRNA protein|S4|DXS306
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: RPS4X
Clonality: Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human