Anti-PROC Mouse Monoclonal Antibody [clone: 3A10]
ABNOH00005624-M01
: Abnova
- Antibody type:Primary
- Antigen name:Protein C
- Antigen symbol:PROC
- Clonality:Monoclonal
- Clone:3A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5624
- Antigen synonyms:APC|THPH4|PC|Coagulation Factor XIV|THPH3|PROC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: PROC
Clonality: Monoclonal
Clone: 3A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human