Anti-PPOX Rabbit Polyclonal Antibody
ABNOH00005498-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Protoporphyrinogen Oxidase
- Antigen symbol:PPOX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5498
- Antigen synonyms:V290M|VP|PPO
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: PPOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human