Anti-PCBP2 Rabbit Polyclonal Antibody
Catalog # ABNOH00005094-D01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Poly(rC) binding protein 2
- Antigen symbol:PCBP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5094
- Antigen synonyms:hnRNP-E2|HNRNPE2|HNRPE2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PCBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human