Anti-PAX3 Mouse Monoclonal Antibody [clone: 4F4]
Catalog # ABNOH00005077-M05
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Paired box 3
- Antigen symbol:PAX3
- Clonality:Monoclonal
- Clone:4F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5077
- Antigen synonyms:CDHS|paired box protein Pax-3|WS3|WS1|paired domain gene HuP2|paired domain gene 3|HUP2|paired box homeotic gene 3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PAX3
Clonality: Monoclonal
Clone: 4F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human