Order Entry
Singapore
ContactUsLinkComponent
Anti-NR4A2 Mouse Monoclonal Antibody [clone: 1D9]
Anti-NR4A2 Mouse Monoclonal Antibody [clone: 1D9]
Catalog # ABNOH00004929-M19
Supplier:  Abnova
undefined
Anti-NR4A2 Mouse Monoclonal Antibody [clone: 1D9]
Catalog # ABNOH00004929-M19
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Nuclear Receptor Subfamily 4, Group A, Member 2
  • Antigen symbol:
    NR4A2
  • Clonality:
    Monoclonal
  • Clone:
    1D9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    4929
  • Antigen synonyms:
    NOT|Nurr1|RNR-1|TINOR|HZF-3|TINUR
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: NR4A2
Clonality: Monoclonal
Clone: 1D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human