Anti-NR4A2 Mouse Monoclonal Antibody [clone: 2A3]
Catalog # ABNOH00004929-M13
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Receptor Subfamily 4, Group A, Member 2
- Antigen symbol:NR4A2
- Clonality:Monoclonal
- Clone:2A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4929
- Antigen synonyms:NOT|Nurr1|RNR-1|TINOR|HZF-3|TINUR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NR4A2
Clonality: Monoclonal
Clone: 2A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human