Anti-NAP1L2 Rabbit Polyclonal Antibody
Catalog # ABNOH00004674-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Nucleosome assembly Protein 1-like 2
- Antigen symbol:NAP1L2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:4674
- Antigen synonyms:MGC26243|NP1L2_HUMAN.|Brain specic gene BPX|Brain specic protein|X linked|BPX|Nucleosome assembly protein 1 like 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NAP1L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human