Anti-MDFI Mouse Monoclonal Antibody [clone: 3B4]
ABNOH00004188-M07
: Abnova
- Antibody type:Primary
- Antigen name:MyoD Family Inhibitor
- Antigen symbol:MDFI
- Clonality:Monoclonal
- Clone:3B4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4188
- Antigen synonyms:Myogenic repressor I-mf|MyoD family inhibitor|MDFI
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: MDFI
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human