Anti-MATN2 Mouse Monoclonal Antibody [clone: 2B8]
Catalog # ABNOH00004147-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Matrilin 2
- Antigen symbol:MATN2
- Clonality:Monoclonal
- Clone:2B8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4147
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MATN2
Clonality: Monoclonal
Clone: 2B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human