Anti-MAN1A1 Rabbit Polyclonal Antibody
Catalog # ABNOH00004121-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Mannosidase alpha class 1A member 1
- Antigen symbol:MAN1A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4121
- Antigen synonyms:MAN9|HUMM9|HUMM3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MAN1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human