Anti-ID2H Mouse Monoclonal Antibody [clone: 2C11]
Catalog # ABNOH00003398-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Inhibitor Of Differentiation 2
- Antigen symbol:ID2H
- Clonality:Monoclonal
- Clone:2C11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3398
- Antigen synonyms:GIG8|ID2A|Cell Growth-Inhibiting Gene 8|Inhibitor Of DNA Binding 2|bHLHb26|Dominant Negative Helix-Loop-Helix Protein|ID2H|DNA-Binding Protein Inhibitor ID2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ID2H
Clonality: Monoclonal
Clone: 2C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human