Order Entry
Singapore
ContactUsLinkComponent
Anti-HK2 Mouse Polyclonal Antibody
Anti-HK2 Mouse Polyclonal Antibody
Catalog # ABNOH00003099-B01P
Supplier:  Abnova
undefined
Anti-HK2 Mouse Polyclonal Antibody
Catalog # ABNOH00003099-B01P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Hexokinase 2
  • Antigen symbol:
    HK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    3099
  • Antigen synonyms:
    HK II|type II|Muscle form hexokinase|Hexokinase
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µg

Specifications

About this item

Type: Primary
Antigen: HK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human