Anti-GTF2A2 Mouse Polyclonal Antibody
Catalog # ABNOH00002958-B02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:General transcription factor 2A, 2 12kDa
- Antigen symbol:GTF2A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2958
- Antigen synonyms:TFIIA|HsT18745|TF2A2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GTF2A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human