Anti-GTF2A2 Mouse Polyclonal Antibody
Catalog # ABNOH00002958-B01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:General transcription factor 2A, 2 12kDa
- Antigen symbol:GTF2A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:2958
- Antigen synonyms:TFIIA|HsT18745|TF2A2
- Storage buffer:No additive
- Sequence:MAYQLYRNTTLGNSLQESLDELIQSQQITPQLALQVLLQFDKAINAALAQRVRNRVNFRGSLNTYRFCDNVWTFVLNDVEFREVTELIKVDKVKIVACDGKNTGSNTTE
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Shipping temperature:Ice
- Immunogen:GTF2A2 (AAH00287, 1 a.a. ~ 109 a.a) full-length human protein.
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: GTF2A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human