Anti-GSTT2 Mouse Monoclonal Antibody [clone: 1C12]
Catalog # ABNOH00002953-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Glutathione S-transferase theta 2
- Antigen symbol:GSTT2
- Clonality:Monoclonal
- Clone:1C12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2953
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GSTT2
Clonality: Monoclonal
Clone: 1C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human