Anti-FHIT Mouse Monoclonal Antibody [clone: 1C3]
Catalog # ABNOH00002272-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fragile Histidine Triad
- Antigen symbol:FHIT
- Clonality:Monoclonal
- Clone:1C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2272
- Antigen synonyms:AP3Aase|FHIT |AP3AaseAP3A hydrolase|FRA3B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: FHIT
Clonality: Monoclonal
Clone: 1C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human