Anti-DNMT3A Rabbit Polyclonal Antibody
ABNOH00001788-D01P
: Abnova
- Antibody type:Primary
- Antigen name:DNA (cytosine-5-)-methyltransferase 3 alpha
- Antigen symbol:DNMT3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1788
- Antigen synonyms:2.1.1.37|DNA (cytosine-5)-methyltransferase 3A|DNA MTase HsaIIIA|M.HsaIIIA|Dnmt3a|DNA methyltransferase HsaIIIA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: DNMT3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human