Order Entry
Singapore
ContactUsLinkComponent
Anti-CCR5 Rabbit Polyclonal Antibody
Anti-CCR5 Rabbit Polyclonal Antibody
  ABNOH00001234-D01P
 :  Abnova
undefined
Anti-CCR5 Rabbit Polyclonal Antibody
  ABNOH00001234-D01P
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Chemokine (C-C motif) receptor 5 (gene/pseudogene)
  • Antigen symbol:
    CCR5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    1234
  • Antigen synonyms:
    IDDM22|CKR5|CD195|CMKBR5|CC-CKR-5|CCCKR5|CCR-5|CKR-5
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: CCR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat