Anti-ART3 Rabbit Polyclonal Antibody
ABNOH00000419-D01
: Abnova
- Antibody type:Primary
- Antigen name:ADP-Ribosyltransferase 3
- Antigen symbol:ART3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:419
- Antigen synonyms:ARTC3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: ART3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human