Anti-ACVRL1 Mouse Monoclonal Antibody [clone: 7H2]
Catalog # ABNOH00000094-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Activin A Receptor Like Type 1
- Antigen symbol:ACVRL1
- Clonality:Monoclonal
- Clone:7H2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:94
- Antigen synonyms:HHT2|SKR3|ALK-1|TSR-I|ALK1|ORW2|ACVRLK1|HHT
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ACVRL1
Clonality: Monoclonal
Clone: 7H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human