Anti-ACVR1B Mouse Monoclonal Antibody [clone: 2D4]
ABNOH00000091-M01
: Abnova
- Antibody type:Primary
- Antigen name:Activin A Receptor Type 1B
- Antigen symbol:ACVR1B
- Clonality:Monoclonal
- Clone:2D4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:91
- Antigen synonyms:Serine/threonine-protein kinase receptor R2|ACTR-IB|Activin receptor-like kinase 4|ALK-4|SKR2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ACVR1B
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human