- Antibody type:Primary
- Antigen name:Thioredoxin Reductase 2
- Antigen symbol:TXNRD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:none
- Gene ID:10587
- Antigen synonyms:SELZ|TR3|TRXR2|TR-BETA|TR
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant human protein purified from E.coli
- Purification:Protein A purified
- Size:100 µl
- Pk:0,1 mL
The mammalian thioredoxin reductases (TrxRs) are a family of selenocysteine-containing pyridine nucleotide-disulfide oxidoreductases. All the mammalian TrxRs are homologous to glutathione reductase with respect to primary structure including the conserved redox catalytic site (-Cys-Val-Asn-Val-Gly-Cys-) but distinctively with a C-terminal extension containing a catalytically active penultimate selenocysteine (SeCys) residue in the conserved sequence(-Gly-Cys-SeCys-Gly). TrxR is homodimeric protein in which each monomer includes an FAD prosthetic group, a NADPH binding site and a redox catalytic site. Electrons are transferred from NADPH via FAD and the active-site disulfide to C-terminal SeCys-containing redox center, which then reduces the substrate like thioredoxin. The members of TrxR family are 55 - 58 kilodalton in molecular size and composed of three isoforms including cytosolic TrxR1, mitochondrial TrxR2, and TrxR3, known as Trx and GSSG reductase (TGR). TrxR plays a key role in protection of cells against oxidative stress and redox-regulatory mechanism of transcription factors and various biological phenomena (1).
Western Blot: 1:2,000;
Type: Primary
Antigen: NR2C1 (nuclear receptor subfamily 2, group C, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse