Order Entry
Singapore
ContactUsLinkComponent
Anti-PRKCG Rabbit Polyclonal Antibody
Anti-PRKCG Rabbit Polyclonal Antibody
Catalog # ABFRLF-PA0174
Supplier:  ABFRONTIER
Anti-PRKCG Rabbit Polyclonal Antibody
Catalog # ABFRLF-PA0174
Supplier:  ABFRONTIER

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase C Gamma
  • Antigen symbol:
    PRKCG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    5582
  • Antigen synonyms:
    PKCG|SCA14|PKCC|PKC-gamma
  • Storage temperature:
    Store for 1 year at –20 °C from date of shipment
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic peptide
  • Purification:
    Protein A column
  • Size:
    100 µl
  • Pk:
    0,1 mL

Specifications

About this item

Protein kinase C (PKC) is a family of serine-threonine kinases that regulate a broad spectrum of cellular functions. The family is composed of nine genes that express structurally related phospholipid-dependent kinases with distinct means of regulation and tissue distribution. Based on their structures and sensitivities to Ca2+ and diacylglycerol (DAG), they have been classified into conventional PKCs (alpha, beta, and gamma), novel PKCs (Delta, Epsilon, Eta, and IPA), and atypical PKCs (Zeta and Lambda/Iota). PKC Gamma is a member of the cPKC subfamily which is activated by Ca2+ and diacylglycerol in the presence of phosphatidylserine. Physiologically, PKC Gamma is activated by a mechanism coupled with receptor-mediated breakdown of inositol phospholipid as other cPKC isotypes. PKC Gamma is expressed solely in the brain and spinal cord and its localization is restricted to neurons. Within the brain, PKC Gamma is the most abundant in the cerebellum, hippocampus and cerebral cortex, where the existence of neuronal plasticity has been demonstrated.

Western Blot: 0.2 ?g/ml; Immunoprecipitation: 2.0 ?g;

Type: Primary
Antigen: PRKCG (protein kinase C, gamma)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat