Order Entry
Singapore
ContactUsLinkComponent
Anti-CSPS Rabbit Polyclonal Antibody
Anti-CSPS Rabbit Polyclonal Antibody
  ABFRLF-PA0081
Anti-CSPS Rabbit Polyclonal Antibody
  ABFRLF-PA0081

 

  • Antibody type:
    Primary
  • Antigen name:
    sulfotransferase family, cytosolic, 1A, phenol-preferring, member 3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    6818
  • Antigen synonyms:
    STM|TL-PST|HAST3|ST1A3|M-PST|HAST|ST1A3/ST1A4|ST1A5
  • Storage temperature:
    Store for 1 year at –20 °C from date of shipment
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant human protein purified from E.coli
  • Purification:
    purified from E.coli
  • Size:
    100 µl
  • Pk:
    0,1 mL

 

 

The human Catecholamine-Sulfating Phenol Sulfotransferase (CSPS) is the only sulfotransferase that catalyses the sulfation of catecholamins, in particular the neurotransmitter dopamine, with high activity. CSPS is required for stimulation by Mn2+ of the sulfating activity and expressed in the human intestine, brain, platelet and other tissues. In the brain it may play a role in regulating the levels of dopamine. It also serves as a detoxifying function in the intestine, where it may detoxify potentially lethal dietary monoamines.

Western Blot: 1:2000; Immunohistochemistry: Paraffin-embedded sections;

Type: Primary
Antigen: CSPS (catecholamine-sulfating phenol sulfotransferase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat