Specifications
- Antibody type:Primary
- Antigen name:tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, beta
- Antigen symbol:YWHAB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Gene ID:7529
- Antigen synonyms:HS1|GW128|KCIP-1|YWHAA|HEL-S-1
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Shipping temperature:Ice
- Immunogen:Recombinant human protein purified from E.coli
- Purification:purified from E.coli
- Size:100 µl
- Pk:0,1 mL
Specifications
About this item
14-3-3, a family of acidic and soluble proteins, highly conserved in amino acid sequences from yeast to mammals, is expressed in all eukaryotic cells. Seven isoforms(β, γ, ε, η, ζ, σ and τ/θ) encoded by seven distinct genes are identified in mammals and forms homo- and hetero- dimeric cup-shaped structures. As 14-3-3 is interacted with more than 100 binding partners, it regulates key proteins involved in various biological processes such as signal trans-duction, cell cycle, transcriptional control, cell proliferation, apoptosis, and ion channel physiology. Most 14-3-3 requires phosphorylation of serine or threonine residues in the target sequence. This protein is abundantly expressed in the brain and has been detected in the cerebrospinal fluid of patients with different neurological disorders.
Western Blot: 1:2000;
Type: Primary
Antigen: 14-3-3 beta (14-3-3 beta)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat