Order Entry
Singapore
ContactUsLinkComponent
Anti-STAT2 Rabbit Polyclonal Antibody
Anti-STAT2 Rabbit Polyclonal Antibody
Catalog # ABFRLF-PA0065
Supplier:  ABFRONTIER
Anti-STAT2 Rabbit Polyclonal Antibody
Catalog # ABFRLF-PA0065
Supplier:  ABFRONTIER

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    signal transducer and activator of transcription 2, 113kDa
  • Antigen symbol:
    STAT2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    6773
  • Antigen synonyms:
    P113|ISGF-3|STAT113
  • Storage temperature:
    Store for 1 year at –20 °C from date of shipment
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic peptide
  • Size:
    100 µl
  • Pk:
    0,1 mL

Specifications

About this item

STAT2 (Signal transducer and activator of transcription 2), 113kDa, is a member of the STAT family of cytoplasmic transcription factors. STAT members generally mediate cytokine, growth factor and hormone receptor signal transduction. STAT2 is a transcription factor critical to the signal transduction pathway of type I interferons (e.g. IFNα). STAT2 resides primarily in the cytoplasm and is tyrosine-phosphorylated after IFNα binds to cell surface receptors. In response to tyrosine phosphorylation STAT2 rapidly localizes to the nucleus and acquires the ability to bind specific DNA targets in association with two other proteins, STAT1 and IFN regulatory factor-9 (IRF-9). STAT2 is phosphorylated at Y689 by receptor-associated Janus kinasses (JAKs) leading to STAT2 dimerization and subsequent translocation to the nucleus to activate gene transcription.

Western Blot: 1:2,000;

Type: Primary
Antigen: STAT2 (signal transducer and activator of transcription 2, 113kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human