Order Entry
Singapore
ContactUsLinkComponent
Anti-CAT Rabbit Polyclonal Antibody
Anti-CAT Rabbit Polyclonal Antibody
Catalog # ABFRLF-PA0060
Supplier:  ABFRONTIER
Anti-CAT Rabbit Polyclonal Antibody
Catalog # ABFRLF-PA0060
Supplier:  ABFRONTIER

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    catalase
  • Antigen symbol:
    CAT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    847
  • Storage temperature:
    Store for 1 year at –20 °C from date of shipment
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant human protein purified from E.coli
  • Purification:
    Ammonium sulfate precipitation
  • Size:
    100 µl
  • Pk:
    0,1 mL

Specifications

About this item

Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kD in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara’s disease (1).

Western Blot: 1:2000; Immunoprecipitation: 1 ?l;

Type: Primary
Antigen: CAT (catalase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat