Specifications
- Antibody type:Primary
- Antigen name:Myeloperoxidase
- Antigen symbol:MPO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Gene ID:4353
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Size:100 µl
- Pk:0,1 mL
Specifications
About this item
Myeloperoxidase is a major neutrophil protein and is also present in monocytes. In neutrophils, it is stored in azurophilic granules and released during phagocytosis. It is a heme enzyme that uses the superoxide and hydrogen peroxide generated by the neutrophil oxidative burst to produce hypochlorous acid and other reactive oxidants.The produced hypochlorous acid reacts with and destroys bacteria. In many inflammatory pathologies, such as cystic fibrosis and rheumatoid arthritis, neutrophils are also causing tissue damage. MPO is thought to be the most promising cardiac marker at the moment. In addition to that MPO is a good inflammatory biomarker for autoimmune, inflammatory diseases and cancer.
Western Blot: 1:2,000;
Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human