Specifications
- Antibody type:Primary
- Antigen name:Methionine sulfoxide reductase A
- Antigen symbol:MSRA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Gene ID:4482
- Antigen synonyms:PMSR
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Shipping temperature:Ice
- Immunogen:Recombinant human protein purified from E.coli
- Purification:purified from E.coli
- Size:100 µl
- Pk:0,1 mL
Specifications
About this item
Methionine sulfoxide reductase (MsrA) reduces methionine sulfoxide (MetO) residues in proteins and free MetO to Methionine (Met). The catalytic activity of MsrA is dependent of bound metal and cofactors but it requires reducing equivalents from either DTT or a thioredoxin-regenerating system. MsrA plays an essential role in protecting cells against oxidative damage. The substrates of MsrA include calmodulin, HIV protease and 1-proteinase-inhibitor (1-3). Recent studies indicate that there is a connection between MsrA and Alzheimer’s disease in mammals (4).
Western Blot: 1:2000;
Type: Primary
Antigen: MSRA (methionine sulfoxide reductase A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat