Order Entry
Singapore
ContactUsLinkComponent
 
Anti-PTPRM Mouse Monoclonal Antibody [clone: T5-AF1A8]
  ABFRLF-MA0354
Anti-PTPRM Mouse Monoclonal Antibody [clone: T5-AF1A8]
  ABFRLF-MA0354

 

  • Antibody type:
    Primary
  • Antigen name:
    Protein tyrosine phosphatase receptor type M
  • Antigen symbol:
    PTPRM
  • Clonality:
    Monoclonal
  • Clone:
    T5-AF1A8
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    5797
  • Antigen synonyms:
    RPTPU|hR-PTPu|RPTPM|PTPRL1|R-PTP-MU
  • Storage temperature:
    Store for 1 year at –20 °C from date of shipmen
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant human protein purified from E.coli
  • Purification:
    Ammonium sulfate precipitation
  • Size:
    100 µl
  • Pk:
    0,1 mL

 

 

Western Blot: 1:1,000; Immunoprecipitation: 1ug;

Type: Primary
Antigen: PTPRM (protein tyrosine phosphatase, receptor type, M)
Clonality: Monoclonal
Clone: T5-AF1A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human