Anti-CTSH Mouse Monoclonal Antibody [clone: AF14D7]
Catalog # ABFRLF-MA0351
Supplier: ABFRONTIER
Specifications
- Antibody type:Primary
- Antigen name:cathepsin H
- Antigen symbol:CTSH
- Clonality:Monoclonal
- Clone:AF14D7
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:none
- Gene ID:1512
- Antigen synonyms:ACC5|minichain|ACC4|CPSB|ACC-4|ACC-5
- Storage temperature:Store for 1 year at –20 °C from date of shipmen
- Shipping temperature:Ice
- Immunogen:Recombinant human protein purified from E.coli (His-Cathepsin H)
- Purification:Ammonium sulfate precipitation
- Size:100 µl
- Pk:0,1 mL
Specifications
About this item
Western Blot: 1:1,000;
Type: Primary
Antigen: CTSH (cathepsin H)
Clonality: Monoclonal
Clone: AF14D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat