- Antibody type:Primary
- Antigen name:lactate dehydrogenase B
- Antigen symbol:LDHB
- Clonality:Monoclonal
- Clone:AF21F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Gene ID:3945
- Antigen synonyms:LDH-H|TRG-5|LDHBD|LDH-B|HEL-S-281
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant human protein purified from E.coli (His-LDHB)
- Purification:Protein G purified
- Size:100 µl
- Pk:0,1 mL
Lactate dehydrogenase (LDH) is the enzyme in the glycolytic pathway that converts pyruvate to lactate with concomitant interconversion of NADH and NAD+. In mammals, the enzyme is encoded by three genes: LDHA (M or muscle form), LDHB (H or heart form), and LDHC (X or testis form). The three human LDHs have 84–89% sequence similarities and 69–75% amino acid identities. There are five different LDH isoenzymes based on the proportion of M and H chains existing in the LDH tetrameric structure.
The LDHA is hypoxia inducible and its expression is directly controlled by the transcriptional activity of the hypoxia inducible factor 1a (HIF1a). LDHB is also known to be upregulated in many cancers.
Western Blot: 0.1 ?g/ml;
Type: Primary
Antigen: LDHB (lactate dehydrogenase B)
Clonality: Monoclonal
Clone: AF21F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat