Order Entry
Singapore
ContactUsLinkComponent
Anti-DUSP4 Mouse Monoclonal Antibody [clone: AFPK7]
Anti-DUSP4 Mouse Monoclonal Antibody [clone: AFPK7]
Catalog # ABFRLF-MA0302
Supplier:  ABFRONTIER
Anti-DUSP4 Mouse Monoclonal Antibody [clone: AFPK7]
Catalog # ABFRLF-MA0302
Supplier:  ABFRONTIER

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Dual specificity phosphatase 4
  • Antigen symbol:
    DUSP4
  • Clonality:
    Monoclonal
  • Clone:
    AFPK7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    1846
  • Antigen synonyms:
    MKP2|HVH2|TYP|MKP-2
  • Storage temperature:
    Store for 1 year at –20 °C from date of shipment
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant human protein purified from E.coli
  • Purification:
    Ammonium sulfate precipitation
  • Size:
    100 µl
  • Pk:
    0,1 mL

Specifications

About this item

There are ten mitogen-activated protein kinase (MAPK) phosphatases (MKPs) that act as negative regulators of MAPK activity in mammalian cells.
MKP2(DUSP4) is a dual threonine/tyrosine phosphatase that specifically dephosphorylates and inactivates MAPKs(ERK, JNK, p38). It contains one Rhodanese domain and one tyrosine-protein phosphatase domain.
MKP2 is expressed in a variety of tissues, and is localized in the nucleus.
Transcription factor E2F-1 acts as a transcriptional regulator of MKP2. E2F-1/MKP2 pathway mediates apoptosis under oxidative stress and that MKP2 suppresses tumor formation.
MKP2 is a novel transcription target of p53 in mediating apoptosis by 10-bp palindrome motif (CTGGCGCCAG) in the MKP2 promoter.

Western Blot: 1:2,000;

Type: Primary
Antigen: DUSP4 (dual specificity phosphatase 4)
Clonality: Monoclonal
Clone: AFPK7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human