- Antibody type:Primary
- Antigen name:Receptor-interacting serine-threonine kinase 2
- Antigen symbol:RIPK2
- Clonality:Monoclonal
- Clone:AF28D3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Gene ID:8767
- Antigen synonyms:CARD3|RICK|CARDIAK|GIG30|CCK|RIP2
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Shipping temperature:Ice
- Immunogen:Recombinant human protein purified from E.coli (His-RIP2)
- Purification:Ammonium sulfate precipitation
- Size:100 µl
- Pk:0,1 mL
The RIP(receptor-interacting protein) family of serine/Threonine kinases(RIP-1,2,3,4,5,6,7) are crucial regulators of cell survival and cell death that can trigger pro-survival, inflammatory and immune responses via the activation of transcription factors(NF-kB and AP-1) and death-inducing programs.
RIP2(also known as RICK, CARDIAK, CCK and Ripk2) transduces signals from receptors of both immune responses. RIP2 carries a CARD at its C-terminal, which is essential for NF-kB activation. RIP2 is a critical downstream mediator of Nod1 and Nod2 signaling. Overexpression of RIP2 results in the activation of, in addition to NF-kB, the MAPKs JNK and ERK2, requiring its kinase activity to activate ERK2, but not JNK.
Western Blot: 1:2,000; Immunoprecipitation: 3 ?l;
Type: Primary
Antigen: RIPK2 (receptor-interacting serine-threonine kinase 2)
Clonality: Monoclonal
Clone: AF28D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat