Order Entry
Singapore
ContactUsLinkComponent
Anti-PTEN Mouse Monoclonal Antibody [clone: 16F3]
Anti-PTEN Mouse Monoclonal Antibody [clone: 16F3]
Catalog # ABFRLF-MA0286
Supplier:  ABFRONTIER
Anti-PTEN Mouse Monoclonal Antibody [clone: 16F3]
Catalog # ABFRLF-MA0286
Supplier:  ABFRONTIER

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    phosphatase and tensin homolog
  • Antigen symbol:
    PTEN
  • Clonality:
    Monoclonal
  • Clone:
    16F3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    5728
  • Antigen synonyms:
    CWS1|BZS|MMAC1|DEC|GLM2|MHAM|10q23del|TEP1|PTEN1
  • Storage temperature:
    Store for 1 year at –20 °C from date of shipment
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant human protein purified from E.coli (His-PTEN)
  • Purification:
    Protein G purified
  • Size:
    100 µl
  • Pk:
    0,1 mL

Specifications

About this item

PTEN acts as a phosphatase to dephosphorylate phosphatidylinositol (3,4,5)-trisphosphate (PtdIns (3,4,5)P3). The product of this enzymatic reaction is PtdIns(4,5)P2. This dephosphorylation is important because it results in inhibition of the AKT signaling pathway.
PTEN is a 403 amino acid protein, and a member of the large PTP (protein tyrosine phosphatase) family. PTEN crystal structure revealed that the N-terminal phosphatase domain is followed by a tightly associated C-terminal C2 domain. These two domains together form a minimal catalytic unit. The phosphatase domain contains the active site which carries out the enzymatic function of the protein, whilst the C2 domain allows PTEN to bind to the phospholipid membrane.
PTEN is one of the most commonly lost tumour suppressors in human cancer, and its deregulation is also implicated in several other diseases. Hereditary mutation of PTEN causes tumor-susceptibility diseases such as Cowden disease.

Western Blot: 0.5 ?g/ml;

Type: Primary
Antigen: PTEN (phosphatase and tensin homolog)
Clonality: Monoclonal
Clone: 16F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat