Specifications
- Antibody type:Primary
- Antigen name:complement component 4A (Rodgers blood group)
- Antigen symbol:C4A
- Clonality:Monoclonal
- Clone:35A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Gene ID:720
- Antigen synonyms:C4A2|CPAMD2|C4A4|C4A3|C4AD|C4S|C4A6|C4|RG|CO4
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Shipping temperature:Ice
- Immunogen:Protein purified from Human plasma
- Purification:Ammonium sulfate precipitation
- Size:100 µl
- Pk:0,1 mL
Specifications
About this item
The complement system is a part of the larger immune system and three biochemical pathways are present: the classical complement pathway, the alternative pathway, and the mannose-binding lectin pathway.
Complement component C4 is an essential component of humoral immune response. In its activated form, C4b becomes a subunit of the C3 convertase, which is an enzymatic complex that activates C3 of the classical and lectin complement activation pathways. The classical pathway is initiated by the activation of the C1-complex (C1q, C1r and C1s) by C1q's binding to antibody-antigen. The C1-complex now binds to and splits C2 and C4 producing C2a and C4b. C4b and C2a bind to form C3-convertase. Production of C3-convertase leads to cleavage of C3 into C3a and C3b and C3b joins with the C3 convertase to make C5 convertase.
Human C4 is the most polymorphic protein of the complement system. Complement C4 exists as two isotypes, C4A (acidic) and C4B (basic). Although the sequence identity is very high, they have different hemolytic activities, covalent affinities to antigens and immune complexes, and serological reactivities. Each C4 contains β chain, α chain, C4a anaphyltoxin, C4b, and γ chain.
C4-deficient mice shows incomplete clearance of microbial attack and C4-deficiency in human shows increased autoimmune diseases.
Western Blot: 1:2,000;
Type: Primary
Antigen: C4B (complement component 4B (Chido blood group))
Clonality: Monoclonal
Clone: 35A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human