Order Entry
Singapore
ContactUsLinkComponent
Anti-SERPINA3 Mouse Monoclonal Antibody [clone: 71A1]
Anti-SERPINA3 Mouse Monoclonal Antibody [clone: 71A1]
Catalog # ABFRLF-MA0166
Supplier:  ABFRONTIER
Anti-SERPINA3 Mouse Monoclonal Antibody [clone: 71A1]
Catalog # ABFRLF-MA0166
Supplier:  ABFRONTIER

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3
  • Antigen symbol:
    SERPINA3
  • Clonality:
    Monoclonal
  • Clone:
    71A1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    12
  • Antigen synonyms:
    AACT|ACT|GIG24|GIG25
  • Storage temperature:
    Store for 1 year at –20 °C from date of shipment
  • Shipping temperature:
    Ice
  • Immunogen:
    Protein purified from Human plasma
  • Purification:
    Ammonium sulfate precipitation
  • Size:
    100 µl
  • Pk:
    0,1 mL

Specifications

About this item

α-1-antichymotrypsin (ACT) is a glycoprotein with a molecular weight of 55~68 kDa and 25% by weight of various sugars, whose heterogeneity results in variation of molecular weight. It inhibits chymotrypsin-like proteinases in vivo and has cytotoxic killer-cell activity in vitro. The protein also has a role as an acute-phase protein and is active in the control of immunologic and inflammatory processes, and as a tumor marker. Plasma ACT levels might increase rapidly more than 5-fold during an acute phase reaction, ACT has been shown to promote Aβ polymerization in vitro and in vivo, and levels of ACT protein in plasma and cerebrospinal fluid from Alzheimer's patients have been found to correlate with progression of dementia.

Western Blot: 1:1,000; Immunohistochemistry: Paraffin-embedded sections;

Type: Primary
Antigen: SERPINA3 (serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3)
Clonality: Monoclonal
Clone: 71A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human