Anti-KLK6 Mouse Monoclonal Antibody [clone: 9A7]
Catalog # ABFRLF-MA0153
Supplier: ABFRONTIER
Specifications
- Antibody type:Primary
- Antigen name:kallikrein-related peptidase 6
- Antigen symbol:KLK6
- Clonality:Monoclonal
- Clone:9A7
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Gene ID:5653
- Antigen synonyms:PRSS18|PRSS9|hK6|Bssp|SP59|Klk7
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Shipping temperature:Ice
- Immunogen:Recombinant human protein purified from E.coli
- Purification:Ammonium sulfate precipitation
- Size:100 µl
- Pk:0,1 mL
Specifications
About this item
Kallikreins(KLKs) belong to the serine protease family of proteolytic enzymes. Neurosin(KLK6), one of these, is atrypsin-like protease dominantly expressed in the human brain but also in a variety of other tissues. As measured by a sensitive quantitative assay in serum and cerebrospinal fluid(CSF), it may have value as a biomarker for diagnosis and monitoring of Alzheimer's disease(AD). It seems to be down regulated in serum and tissues of AD patients and may be involved in amyloid metabolism.
Western Blot: 1:1,000;
Type: Primary
Antigen: KLK6 (kallikrein-related peptidase 6)
Clonality: Monoclonal
Clone: 9A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human