Order Entry
Singapore
ContactUsLinkComponent
Anti-Fibronectin Mouse Monoclonal Antibody [clone: 23C9]
Anti-Fibronectin Mouse Monoclonal Antibody [clone: 23C9]
Catalog # ABFRLF-MA0142
Supplier:  ABFRONTIER
Anti-Fibronectin Mouse Monoclonal Antibody [clone: 23C9]
Catalog # ABFRLF-MA0142
Supplier:  ABFRONTIER

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fibronectin
  • Antigen symbol:
    FN1
  • Clonality:
    Monoclonal
  • Clone:
    23C9
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    none
  • Gene ID:
    2335
  • Antigen synonyms:
    ED-B|FNZ|MSF|CIG|GFND2|LETS|GFND|FN|FINC
  • Storage temperature:
    Store for 1 year at –20 °C from date of shipment
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Fibronectin protein purified from Human plasma
  • Purification:
    Protein G purified
  • Size:
    100 µl
  • Pk:
    0,1 mL

Specifications

About this item

Fibronectin (FN) exists in two main forms: 1) as a soluble glycoprotein in blood plasma (plasma FN), and 2) as an insoluble glycoprotein in tissue extracellular matrices (cellular FN). Many different cell types synthesize fibronectin and secrete it as a disulfide-bonded dimer composed of 230–270 kDa subunits. FN is one of the largest multi-domain proteins that interact with a variety of macromolecules like heparin, collagen /gelatin, and fibrin. FN is involved in many cellular processes, including tissue repair, embryogenesis, blood clotting, and cell migration/ adhesion and so can be used as a therapeutic agent for wound healing. In addition, its age-dependent increase in plasma and tissues may be accompanied in pathological states, especially in tumor growth, by its proteolytic breakdown.

Western Blot: 1.0ug/ml; Immunoprecipitation: 2.0ug;

Type: Primary
Antigen: Fibronectin (fibronectin)
Clonality: Monoclonal
Clone: 23C9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human