Specifications
- Antibody type:Primary
- Antigen name:complement component 3
- Antigen symbol:C3
- Clonality:Monoclonal
- Clone:10A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Gene ID:718
- Antigen synonyms:C3b|AHUS5|ARMD9|C3a|ASP|CPAMD1|HEL-S-62p
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Complement C3 protein purified from Human plasma
- Purification:Protein G purified
- Size:100 µl
- Pk:0,1 mL
Specifications
About this item
Human Complement C3 (C3) is synthesized as a single-chain pro-molecule (185 kDa) that then suffers several post-translational modifications. Before being secreted as a mature protein, C3 is split into -chain (645 residues and 70 kDa) and chain (992 residues and 115 kDa) and forms a rare internal thioester bond. C3 plays a central role in the activation of all the three pathways of complement activation i.e. the classical, alternative, and lectin pathway. As C3 is the major complement component and participates in several stages of the immune response, its deficiency generally associated with higher susceptibility to severe bacterial infections and in some cases with autoimmune diseases such as systemic lupus erythematosus.
Western Blot: 0.5 ?g/ml;
Type: Primary
Antigen: C3 (complement component 3)
Clonality: Monoclonal
Clone: 10A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human