- Antibody type:Primary
- Antigen name:Heat shock 70kDA protein 1A
- Antigen symbol:HSPA1A
- Clonality:Monoclonal
- Clone:16A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Gene ID:3303
- Antigen synonyms:HSPA1|HSP70-1A|HSP72|HSP70-1|HSP70.1|HEL-S-103|HSP70I
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Shipping temperature:Ice
- Immunogen:Recombinant Human HSP70 protein purified E.coli
- Purification:Ammonium sulfate precipitation
- Size:100 µl
- Pk:0,1 mL
The Heat shock protein 70(HSP70) family was found in many intracellular compartments. Members of this protein occur in chloroplasts, endoplasmic reticulum, mitochondria, and cytosol. These proteins are induced by a variety of biological stresses, including heat stress, in every organism. HSP70 serves a variety of roles: 1) It acts as molecular chaperones facilitating the assembly of multi-protein complexes, 2) It participates in the translocation of polypeptides across cell membranes and to the nucleus 3) It aids in the proper folding of nascent polypeptide chains. HSP70 is mitochondrial import machinery and plays key roles in the cytosolic endoplasmic reticulum. Recently, extracellular localized HSP have been found to play key roles in the induction of a cellular immune response.
Western Blot: 1:2000; Immunoprecipitation: 1-2 ?l; Immunohistochemistry: Paraffin-embedded sections;
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: Monoclonal
Clone: 16A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Gamma
Reactivity: Human