Anti-PRDX-SO3 Mouse Monoclonal Antibody [clone: 10A1]
Catalog # ABFRLF-MA0088
Supplier: ABFRONTIER
Specifications
- Cross adsorption:none
- Antigen synonyms:MSP23|PAGA|PAG|NKEF-A|PRXI|NKEFA|PRX1|TDPX2|PAGB
- Storage temperature:Store for 1 year at –20 °C from date of shipment
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Sulfonylated peptide(KLH coupled) corresponding to the active site sequence common to mammalian Prx I to IV
- Purification:Protein G purified
- Size:100 µl
- Cat. no.:ABFRLF-MA0088
- Pk:0,1 mL