Order Entry
Singapore
ContactUsLinkComponent
Anti-SOD4 Mouse Monoclonal Antibody [clone: 3A1]
Anti-SOD4 Mouse Monoclonal Antibody [clone: 3A1]
Catalog # ABFRLF-MA0042
Supplier:  ABFRONTIER
Anti-SOD4 Mouse Monoclonal Antibody [clone: 3A1]
Catalog # ABFRLF-MA0042
Supplier:  ABFRONTIER

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    copper chaperone for superoxide dismutase
  • Antigen symbol:
    CCS
  • Clonality:
    Monoclonal
  • Clone:
    3A1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Gene ID:
    9973
  • Storage temperature:
    Store for 1 year at –20 °C from date of shipment
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant human protein purified from E.coli
  • Purification:
    Protein G purified
  • Size:
    100 µl
  • Pk:
    0,1 mL

Specifications

About this item

Superoxide dismutase (SOD) is an antioxidant enzyme involved in the defense system against reactive oxygen species (ROS). SOD catalyzes the dismutation reaction of superoxide radical anion (O2-) to hydrogen peroxide, which is then catalyzed to innocuous O2 and H2O by glutathione peroxidase and catalase. Several classes of SOD have been identified. These include intracellular copper, zinc SOD (Cu, Zn-SOD/SOD-1), mitochondrial manganese SOD (Mn-SOD/SOD-2) and extracellular Cu, Zn-SOD (EC-SOD/SOD-3) (1). SOD1 is found in all eukaryotic species as a homodimeric 32 kDa enzyme containing one each of Cu and Zn ion per subunit (2). The manganese containing 80 kDa tetrameric enzyme SOD2, is located in the mitochondrial matrix in close proximity to a primary endogenous source of superoxide, the mitochondrial respiratory chain (3). SOD3 is a heparin-binding multimer of disulfide-linked dimers, primarily expressed in human lungs, vessel walls and airways (4). SOD4 is a copper chaperone for superoxide dismutase (CCS), which specifically delivers Cu to copper/zinc superoxide dismutase. CCS may activate copper/zinc superoxide dismutase through direct insertion of the Cu cofactor.

Western Blot: 0.2 ?g/ml; Immunoprecipitation: 2.0 ?g; Immunohistochemistry: Paraffin-embedded sections;

Type: Primary
Antigen: SOD4 (superoxide dismutase 4)
Clonality: Monoclonal
Clone: 3A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human