Neptune® Microcentrifuge Tubes
BITX63300641
: BIOTIX
- Packed:250/pack
- Sterile:+
- Type:with attached lid
- Version:graduated, conical bottom, sterile
- Capacity:2,0 ml
- Colour:Colourless
- Seq:03
- Pk:250
PP, translucent.
- Graduated, with flat cap
- DNase, RNase and endotoxin free
- Centrifugation up to 20000×g
- Autoclavable